missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Mucin 5AC, Clone: SPM297, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Mucin 5AC
Clone SPM297
Concentration 1mg/mL
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Gene Accession No. P98088
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23475801 Novus Biologicals Canada
NBP23475801MG
0.1mg Each for $384.00
NBP23475802 Novus Biologicals Canada
NBP23475802MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Mucin 5AC
1mg/mL
P98088
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM297
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
4586
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot