Please login to your online account to display your discounted pricing

Mouse anti-Mucin 5AC, Clone: SPM297, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475802

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Antigen Mucin 5AC
Format Purified
Primary or Secondary Primary
Gene ID (Entrez) 4586
Isotype IgG1, κ
Concentration 1mg/mL
Conjugate Unlabeled
Quantity 0.2mg
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Test Specificity This MAb recognizes the peptide core of gastric mucin M1 (>1,000kDa) (recently identified as Mucin 5AC). Does not react to Bovine.
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Purification Method Purified
Gene Accession No. P98088
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Clone SPM297