Please login to your online account to display your discounted pricing

Mouse anti-Mucin 5AC, Clone: SPM297, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Antigen Mucin 5AC
Isotype IgG1, κ
Format Purified
Quantity 0.1mg
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Test Specificity This MAb recognizes the peptide core of gastric mucin M1 (>1,000kDa) (recently identified as Mucin 5AC). Does not react to Bovine.
Reconstitution Protein A purified
Clone SPM297
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Regulatory Status RUO
Gene ID (Entrez) 4586
Purification Method Purified
Concentration 1mg/mL
Gene Accession No. P98088