Please login to your online account to display your discounted pricing

Mouse anti-MUC2, Clone: SPM296, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen MUC2
Clone SPM296
Concentration 0.2mg/mL
Immunogen A synthetic peptide of 29 amino acids KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein
Gene Accession No. Q02817
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23298410

 
novus biologicals canada
NBP23298410ML
1.0mL Each for $429.00

NBP23298401

 
novus biologicals canada
NBP23298401ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen MUC2
Clone SPM296
Concentration 0.2mg/mL
Immunogen A synthetic peptide of 29 amino acids KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein
Gene Accession No. Q02817
Gene ID (Entrez) 4583
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting