Please login to your online account to display your discounted pricing

Mouse anti-MUC2, Clone: SPM296, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23298410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23298410

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen MUC2
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q02817
Immunogen A synthetic peptide of 29 amino acids KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4583
Reconstitution Protein A purified
Clone SPM296
Conjugate Unlabeled
Host Species Mouse
Concentration 0.2mg/mL