Please login to your online account to display your discounted pricing

Mouse anti-MUC2, Clone: SPM296, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475701

  • / Each
 

Description & Specifications

Specifications

Antigen MUC2
Format Purified
Gene Accession No. Q02817
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide of 29 amino acids
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4583
Test Specificity Recognizes a single glycoprotein of 520kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3.
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Reconstitution Protein A purified
Clone SPM296