Please login to your online account to display your discounted pricing

Mouse anti-MUC2, Clone: SPM296, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475702

  • / Each
 

Description & Specifications

Specifications

Antigen MUC2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM296
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q02817
Host Species Murine
Immunogen A synthetic peptide of 29 amino acids
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4583
Test Specificity Recognizes a single glycoprotein of 520kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3.
Reconstitution Protein A purified
Target Species Human