Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM533, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen MUC-1
Clone SPM533
Concentration 0.2mg/mL
Immunogen Delipidated extract of human milk fat globule membranes
Gene Accession No. P15941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23287810

 
novus biologicals canada
NBP23287810ML
1.0mL Each for $429.00

NBP23287801

 
novus biologicals canada
NBP23287801ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen MUC-1
Clone SPM533
Concentration 0.2mg/mL
Immunogen Delipidated extract of human milk fat globule membranes
Gene Accession No. P15941
Gene ID (Entrez) 4582
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting