Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM533, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23287810ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23287810

  • / Each
 

Description & Specifications

Specifications

Antigen MUC-1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM533
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P15941
Host Species Mouse
Immunogen Delipidated extract of human milk fat globule membranes
Isotype IgG2a, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4582
Reconstitution Protein A purified