Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM533, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23287801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23287801

  • / Each
 

Description & Specifications

Specifications

Antigen MUC-1
Format Purified
Gene Accession No. P15941
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Delipidated extract of human milk fat globule membranes
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4582
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Host Species Mouse
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM533