Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM533, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Primary or Secondary Primary
Cross Reactivity Human
Antigen MUC-1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P15941
Immunogen Delipidated extract of human milk fat globule membranes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4582
Test Specificity In Western blotting, it recognizes proteins in MW range of 265-400kDa, identified as different glycoforms of EMA.
Clone SPM533
Conjugate Unlabeled
Host Species Mouse
Purification Method Purified
Quantity 0.1mg
Reconstitution Protein A purified
Concentration 1mg/mL