Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM533, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478202

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Quantity 0.2mg
Immunogen Delipidated extract of human milk fat globule membranes
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Host Species Mouse
Test Specificity In Western blotting, it recognizes proteins in MW range of 265-400kDa, identified as different glycoforms of EMA.
Gene Accession No. P15941
Concentration 1mg/mL
Reconstitution Protein A purified
Gene ID (Entrez) 4582
Monoclonal or Polyclonal Monoclonal
Clone SPM533
Primary or Secondary Primary
Format Purified
Antigen MUC-1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.