missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM132, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen MUC-1
Clone SPM132
Concentration 1mg/mL
Immunogen Human milk fat globule membranes
Gene Accession No. P15941
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23473701 Novus Biologicals Canada
NBP23473701MG
0.1mg Each for $384.00
NBP23473702 Novus Biologicals Canada
NBP23473702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

MUC-1
1mg/mL
P15941
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM132
Human milk fat globule membranes
4582
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot