Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM132, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473702

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human milk fat globule membranes
Cross Reactivity Human
Antigen MUC-1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4582
Test Specificity In Western blotting, it recognizes proteins in MW range of 265-400kDa, identified as different glycoforms of EMA. This MAb reacts with the DTRP epitope in the tandem repeats.
Reconstitution Protein A purified
Clone SPM132
Concentration 1mg/mL
Conjugate Unlabeled
Gene Accession No. P15941
Host Species Mouse