Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: SPM132, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473701

  • / Each
 

Description & Specifications

Specifications

Antigen MUC-1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human milk fat globule membranes
Isotype IgG1, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4582
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P15941
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In Western blotting, it recognizes proteins in MW range of 265-400kDa, identified as different glycoforms of EMA. This MAb reacts with the DTRP epitope in the tandem repeats.
Clone SPM132
Concentration 1mg/mL