Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: GP1.4 + E29, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen MUC-1
Clone GP1.4 + E29
Concentration 0.2mg/mL
Immunogen Human milk fat globule membranes (GP1.4); Delipidated extract of human milk fat globule membranes (E29)
Gene Accession No. P15941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23427601

 
novus biologicals canada
NBP23427601ML
0.1mL Each for $139.00

NBP23427605

 
novus biologicals canada
NBP23427605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen MUC-1
Clone GP1.4 + E29
Concentration 0.2mg/mL
Immunogen Human milk fat globule membranes (GP1.4); Delipidated extract of human milk fat globule membranes (E29)
Gene Accession No. P15941
Gene ID (Entrez) 4582
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting