Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: GP1.4 + E29, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427605

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Gene Accession No. P15941
Host Species Mouse
Immunogen Human milk fat globule membranes (GP1.4); Delipidated extract of human milk fat globule membranes (E29)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4582
Concentration 0.2mg/mL
Conjugate Unlabeled
Antigen MUC-1
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone GP1.4 + E29
Cross Reactivity Human