Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: GP1.4 + E29, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Clone GP1.4 + E29
Concentration 1mg/mL
Immunogen Human milk fat globule membranes (GP1.4); Delipidated extract of human milk fat globule membranes (E29)
Format Purified
Regulatory Status RUO
Cross Reactivity Human
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4582
Isotype IgG1, κ
Antigen MUC-1
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Gene Accession No. P15941
Quantity 0.1mg