Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: E29, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen MUC-1
Clone E29
Concentration 1mg/mL
Immunogen Delipidated extract of human milk fat globule membranes
Gene Accession No. P15941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23461002

 
novus biologicals canada
NBP23461002MG
0.2mg Each for $527.00

NBP23461001

 
novus biologicals canada
NBP23461001MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen MUC-1
Clone E29
Concentration 1mg/mL
Immunogen Delipidated extract of human milk fat globule membranes
Gene Accession No. P15941
Gene ID (Entrez) 4582
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot