Please login to your online account to display your discounted pricing

Mouse anti-MUC-1, Clone: E29, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461001

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Antigen MUC-1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Delipidated extract of human milk fat globule membranes
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Format Purified
Isotype IgG2a, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 4582
Reconstitution Protein A purified
Clone E29
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P15941