Please login to your online account to display your discounted pricing

Mouse anti-MSH6, Clone: SPM525, Novus Biologicals™

$307.00 - $518.93

Specifications

Antigen MSH6
Clone SPM525
Immunogen A synthetic peptide corresponding to internal region of human MSH6 protein.
Gene ID (Entrez) 2956
Formulation 10mM PBS, pH 7.4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13615301

 
novus biologicals canada
NBP13615301ML
0.1mL Each for $307.00

NBP13615310

 
novus biologicals canada
NBP13615310ML
1mL Each for $518.93
Description & Specifications

Specifications

Antigen MSH6
Clone SPM525
Immunogen A synthetic peptide corresponding to internal region of human MSH6 protein.
Gene ID (Entrez) 2956
Formulation 10mM PBS, pH 7.4
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)