Please login to your online account to display your discounted pricing

Mouse anti-MSH6, Clone: SPM525, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13615370ML

 View all options for this product


For Research Use Only

Catalog No. NBP13615370


Description & Specifications


Antigen MSH6
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SPM525
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 10mM PBS, pH 7.4
Host Species Mouse
Immunogen A synthetic peptide corresponding to internal region of human MSH6 protein.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 7mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2956
Test Specificity This product is specific for MSH6