Please login to your online account to display your discounted pricing

Mouse anti-MSH6, Clone: SPM525, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13615310ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13615310

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Isotype IgG1, κ
Immunogen A synthetic peptide corresponding to internal region of human MSH6 protein.
Gene ID (Entrez) 2956
Format Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Human
Quantity 1mL
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Antigen MSH6
Formulation 10mM PBS, pH 7.4
Test Specificity This product is specific for MSH6
Primary or Secondary Primary
Clone SPM525
Purification Method Affinity Purified
Host Species Mouse