Please login to your online account to display your discounted pricing

Mouse anti-MSH6, Clone: SPM525, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13615301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13615301

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation 10mM PBS, pH 7.4
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Isotype IgG1, κ
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2956
Clone SPM525
Cross Reactivity Human
Antigen MSH6
Host Species Mouse
Immunogen A synthetic peptide corresponding to internal region of human MSH6 protein.
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Test Specificity This product is specific for MSH6
Conjugate Unlabeled