Please login to your online account to display your discounted pricing

Mouse anti-MS4A1/CD20, Clone: B9E9, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen MS4A1/CD20
Clone B9E9
Concentration 1mg/mL
Immunogen Lymphoblastoid cell line Daudi
Gene Accession No. P11836
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23455901

 
novus biologicals canada
NBP23455901MG
0.1mg Each for $384.00

NBP23455902

 
novus biologicals canada
NBP23455902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen MS4A1/CD20
Clone B9E9
Concentration 1mg/mL
Immunogen Lymphoblastoid cell line Daudi
Gene Accession No. P11836
Gene ID (Entrez) 931
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation