Please login to your online account to display your discounted pricing

Mouse anti-MS4A1/CD20, Clone: 2H7, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen MS4A1/CD20
Clone 2H7
Concentration 0.5mg/mL
Immunogen Made to Human CD20
Gene ID (Entrez) 931
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14366701

 
novus biologicals canada
NBP14366701MG
0.1mg Each for $252.00

NBP14366703

 
novus biologicals canada
NBP1436670025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen MS4A1/CD20
Clone 2H7
Concentration 0.5mg/mL
Immunogen Made to Human CD20
Gene ID (Entrez) 931
Formulation PBS [pH 7.2]. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2b, κ
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)