Please login to your online account to display your discounted pricing

Mouse anti-Moesin, Clone: SPM562, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Moesin
Clone SPM562
Concentration 0.2mg/mL
Immunogen Recombinant human Moesin protein
Gene Accession No. P26038
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23287601

 
novus biologicals canada
NBP23287601ML
0.1mL Each for $139.00

NBP23287610

 
novus biologicals canada
NBP23287610ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Moesin
Clone SPM562
Concentration 0.2mg/mL
Immunogen Recombinant human Moesin protein
Gene Accession No. P26038
Gene ID (Entrez) 4478
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting