Please login to your online account to display your discounted pricing

Mouse anti-Moesin, Clone: SPM562, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23287601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23287601

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Antigen Moesin
Formulation PBS with 0.05% BSA.
Gene Accession No. P26038
Immunogen Recombinant human Moesin protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4478
Test Specificity Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Clone SPM562
Concentration 0.2mg/mL
Regulatory Status RUO
Reconstitution Protein A purified