missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Moesin, Clone: MSN491, Azide Free, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23468501MG

 View more versions of this product

Catalog No. NBP23468501


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Moesin
MSN491
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG1, κ
0.1mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Human
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
1mg/mL
Purified
P26038
Recombinant human Moesin protein
Purified
RUO
Primary
4478
Protein A purified