Please login to your online account to display your discounted pricing

Mouse anti-Moesin, Clone: MSN491, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468502

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Primary or Secondary Primary
Gene ID (Entrez) 4478
Conjugate Unlabeled
Antigen Moesin
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P26038
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Concentration 1mg/mL
Immunogen Recombinant human Moesin protein
Isotype IgG1, κ
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone MSN491
Cross Reactivity Human