Please login to your online account to display your discounted pricing

Mouse anti-MMP-12, Clone: 705D10.11, HRP, Novus Biologicals™

$439.00 - $439.00

Specifications

Antigen MMP-12
Clone 705D10.11
Concentration 0.5mg/mL
Immunogen MMP-12/HME
Gene ID (Entrez) 4321
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

DDX0282HRP1

 
novus biologicals canada
DDX0282HRPO100
0.1mg Each for $439.00

DDX0282HRP5

 
novus biologicals canada
DDX0282HRPO100005MG
0.05mg Each for $439.00
Description & Specifications

Specifications

Antigen MMP-12
Clone 705D10.11
Concentration 0.5mg/mL
Immunogen MMP-12/HME
Gene ID (Entrez) 4321
Formulation Tris-NaCl pH 8
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate HRP
Format Purified
Purification Method Ion Exchange Chromatography
Applications ELISA