missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-MMP-12, Clone: 701E4.03, HRP, Novus Biologicals™

$439.00 - $439.00

Specifications

Antigen MMP-12
Clone 701E4.03
Concentration 0.5mg/mL
Immunogen MMP-12/HME
Gene ID (Entrez) 4321
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
DDX0284HRP1 Novus Biologicals Canada
DDX0284HRPO100
0.1mg Each for $439.00
DDX0284HRP5 Novus Biologicals Canada
DDX0284HRPO100005MG
0.05mg Each for $439.00
Specifications Description & Specifications

Specifications

MMP-12
0.5mg/mL
4321
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA
701E4.03
MMP-12/HME
Tris-NaCl pH 8
Primary
Murine
HRP
Ion Exchange Chromatography