Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: SPM198, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23298110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23298110

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Conjugate Unlabeled
Cross Reactivity Human
Antigen Mitochondria
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Semi-purified mitochondrial preparation
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Does not react to Mouse and Rat.
Reconstitution Protein A purified
Clone SPM198
Concentration 0.2mg/mL