Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: SPM198, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474502

  • / Each
 

Description & Specifications

Specifications

Format Purified
Cross Reactivity Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Mitochondria
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Isotype IgG1, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Clone SPM198
Purification Method Purified
Concentration 1mg/mL
Host Species Mouse
Reconstitution Protein A purified
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Immunogen Semi-purified mitochondrial preparation
Quantity 0.2mg
Test Specificity MAb recognizes a 60kDa antigen associated with the mitochondria in human cells. It reacts specifically with human cells, including neurons and embryonic stem cells. Does not react with Mouse or Rat.