Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: SPM198, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Reconstitution Protein A purified
Isotype IgG1, κ
Regulatory Status RUO
Quantity 0.1mg
Antigen Mitochondria
Test Specificity MAb recognizes a 60kDa antigen associated with the mitochondria in human cells. It reacts specifically with human cells, including neurons and embryonic stem cells. Does not react with Mouse or Rat.
Concentration 1mg/mL
Primary or Secondary Primary
Immunogen Semi-purified mitochondrial preparation
Clone SPM198