Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: MTC719, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468701

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen Mitochondria
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Mitochondrial fraction of HeLa cells
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity Does not react with insects or bacteria.
Cross Reactivity Human
Purification Method Purified
Reconstitution Protein A purified
Clone MTC719
Concentration 1mg/mL