Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: MTC02, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468601

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Primary or Secondary Primary
Format Purified
Quantity 0.1mg
Purification Method Purified
Cross Reactivity Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Semi-purified mitochondrial preparation
Reconstitution Protein A purified
Antigen Mitochondria
Conjugate Unlabeled
Isotype IgG1, κ
Clone MTC02
Concentration 1mg/mL
Test Specificity Does not react to Mouse or Rat.