Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: 113-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Mitochondria
Clone 113-1
Concentration 1mg/mL
Immunogen Semi-purified mitochondrial preparation
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23451701

 
novus biologicals canada
NBP23451701MG
0.1mg Each for $384.00

NBP23451702

 
novus biologicals canada
NBP23451702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Mitochondria
Clone 113-1
Concentration 1mg/mL
Immunogen Semi-purified mitochondrial preparation
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot