Please login to your online account to display your discounted pricing

Mouse anti-Mitochondria, Clone: 113-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451702

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Semi-purified mitochondrial preparation
Format Purified
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity Does not react to Mouse or Rat.
Cross Reactivity Human
Antigen Mitochondria
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone 113-1
Concentration 1mg/mL
Conjugate Unlabeled