Please login to your online account to display your discounted pricing

Mouse anti-MHC Class II, Clone: 14-4-4S, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen MHC Class II
Clone 14-4-4S
Immunogen The immunogen for this antibody was MHC class II.
Gene ID (Entrez) 3108
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14350701

 
novus biologicals canada
NBP14350701MG
0.1mg Each for $329.00

NBP14350705

 
novus biologicals canada
NBP143507005MG
0.05mg Each for $252.00

NBP1435075

 
novus biologicals canada
NBP14350705MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen MHC Class II
Clone 14-4-4S
Immunogen The immunogen for this antibody was MHC class II.
Gene ID (Entrez) 3108
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry