Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: SF1-1.1.1, APC, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen MHC Class I
Clone SF1-1.1.1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14298801

 
novus biologicals canada
NBP14298801MG
0.1mg Each for $362.00

NBP14298803

 
novus biologicals canada
NBP1429880025MG
0.025mg Each for $197.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone SF1-1.1.1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry