Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: AF6-88.5.5.3, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen MHC Class I
Clone AF6-88.5.5.3
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14351301

 
novus biologicals canada
NBP14351301MG
0.1mg Each for $329.00

NBP14351303

 
novus biologicals canada
NBP1435130025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone AF6-88.5.5.3
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry