Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: A4, Novus Biologicals™

$230.00 - $329.00

Specifications

Antigen MHC Class I
Clone A4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was HLA MHC Class I free chain without B2 microglobulin.
Gene ID (Entrez) 3133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14312201

 
novus biologicals canada
NBP14312201MG
0.1mg Each for $329.00

NBP14312203

 
novus biologicals canada
NBP1431220025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone A4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was HLA MHC Class I free chain without B2 microglobulin.
Gene ID (Entrez) 3133
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation