Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: A4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431220025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14312203

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Primary or Secondary Primary
Antigen MHC Class I
Conjugate Unlabeled
Gene ID (Entrez) 3133
Regulatory Status RUO
Clone A4
Cross Reactivity Human
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Immunogen The immunogen for this antibody was HLA MHC Class I free chain without B2 microglobulin.
Host Species Mouse
Formulation PBS [pH 7.2]
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Purification Method Affinity Purified
Quantity 0.025mg