Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 3D12HLA-E, Novus Biologicals™

$285.00 - $329.00

Specifications

Antigen MHC Class I
Clone 3D12HLA-E
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was HLAE.
Gene ID (Entrez) 3133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14312401

 
novus biologicals canada
NBP14312401MG
0.1mg Each for $329.00

NBP14312403

 
novus biologicals canada
NBP1431240025MG
0.025mg Each for $285.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone 3D12HLA-E
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was HLAE.
Gene ID (Entrez) 3133
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting