Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 3D12HLA-E, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14312401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14312401

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Format Affinity Purified
Antigen MHC Class I
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Clone 3D12HLA-E
Primary or Secondary Primary
Gene ID (Entrez) 3133
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation PBS [pH 7.2]
Concentration 0.2mg/mL
Host Species Mouse
Quantity 0.1mg
Isotype IgG1
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Immunogen The immunogen for this antibody was HLAE.