Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, PE, Novus Biologicals™

$246.00 - $493.00

Specifications

Antigen MHC Class I
Clone 28-14-8
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14373001

 
novus biologicals canada
NBP14373001MG
0.1mg Each for $354.00

NBP14373005

 
novus biologicals canada
NBP143730005MG
0.05mg Each for $246.00

NBP14373002

 
novus biologicals canada
NBP14373002MG
0.2mg Each for $493.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone 28-14-8
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry