Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14373001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14373001

  • / Each
 

Description & Specifications

Specifications

Conjugate PE
Antigen MHC Class I
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was MHC Class I.
Isotype IgG2a, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3133
Clone 28-14-8
Cross Reactivity Mouse
Applications Flow Cytometry
Quantity 0.1mg