Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, Novus Biologicals™

$274.00 - $362.00

Specifications

Antigen MHC Class I
Clone 28-14-8
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14325501

 
novus biologicals canada
NBP14325501MG
0.1mg Each for $274.00

NBP1432555

 
novus biologicals canada
NBP14325505MG
0.5mg Each for $362.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone 28-14-8
Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry