Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432555

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Immunogen The immunogen for this antibody was MHC Class I.
Quantity 0.5mg
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Clone 28-14-8
Conjugate Unlabeled
Isotype IgG2a, κ
Antigen MHC Class I
Format Affinity Purified
Purification Method Affinity Purified
Primary or Secondary Primary
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Gene ID (Entrez) 3133
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Mouse